siRNA Details
Gene Symbol: Zcchc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106658.2 | NP_001100128.1 | 2 | 92 | ||
GenBank mRNA | BC161967.1 | - | 2 | 92 | 9028474 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zcchc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106658.2 | NP_001100128.1 | 2 | 92 | ||
GenBank mRNA | BC161967.1 | - | 2 | 92 | 9028474 |