siRNA Details
Gene Symbol: Hnrnpm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109911.1 | NP_001103381.1 | 14 | 1317 | ||
NM_053876.2 | NP_446328.2 | 15 | 1200 | |||
XM_006241116.2 | XP_006241178.1 | 14 | 2507 | |||
XM_008765198.1 | XP_008763420.1 | 12 | 1047 | |||
GenBank mRNA | BC088317.1 | - | 10 | 828 | 7325202 | |
U32577.2 | - | 15 | 1200 |
