siRNA Details
Gene Symbol: Mettl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163164.1 | NP_001156636.1 | 5 | 859 | ||
XM_006230154.2 | XP_006230216.1 | 5 | 1088 | |||
XM_008759659.1 | XP_008757881.1 | 5 | 748 | |||
GenBank mRNA | FJ889987.1 | - | 5 | 858 | ||
HQ898855.1 | - | 5 | 856 |
