siRNA Details
Gene Symbol: LOC683722 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101008.1 | NP_001094478.1 | 5 | 469 | ||
XM_008758772.1 | XP_008756994.1 | 5 | 880 | |||
XM_008758773.1 | XP_008756995.1 | 5 | 877 | |||
XM_008758774.1 | XP_008756996.1 | 5 | 574 | |||
GenBank mRNA | BC098026.1 | - | 5 | 469 | 7455942 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_201230, NM_001197046, BC080717, BB615739, AK050389, and others.
Gene Ontology Categories:
