siRNA Details
Gene Symbol: Lpgat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109376.2 | NP_001102846.1 | 2 | 364 | ||
XM_008769875.1 | XP_008768097.1 | 3 | 422 |

More Information
Other Species Targeted

Gene Ontology Categories:
