siRNA Details
Gene Symbol: Ubxn2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109482.1 | NP_001102952.1 | 4 | 410 | ||
XM_006239861.2 | XP_006239923.1 | 4 | 469 |

More Information
Other Species Targeted

Gene Ontology Categories:
