siRNA Details
Gene Symbol: Ndufab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106294.1 | NP_001099764.1 | 2 | 305 | ||
XM_006230188.1 | XP_006230250.1 | 2 | 337 | |||
GenBank mRNA | FQ212307.1 | - | 2 | 304 | ||
FQ212475.1 | - | 2 | 342 | |||
FQ216942.1 | - | 2 | 282 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
