siRNA Details
Gene Symbol: Eif4g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248581.2 | XP_006248643.1 | 25 | 4074 | ||
XM_006248582.1 | XP_006248644.1 | 25 | 4063 | |||
XM_006248583.1 | XP_006248645.1 | 25 | 4054 | |||
XM_006248584.2 | XP_006248646.1 | 26 | 4141 | |||
XM_006248585.2 | XP_006248647.1 | 25 | 4082 | |||
XM_008768835.1 | XP_008767057.1 | 26 | 4138 | |||
GenBank mRNA | FQ217072.1 | - | 3 | 452 |
Gene Symbol: LOC100912571 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248555.1 | XP_006248617.1 | 865 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
