siRNA Details
Gene Symbol: Tmem52b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109560.1 | NP_001103030.1 | 4 | 814 | ||
XM_006237102.2 | XP_006237164.1 | 5 | 751 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
