siRNA Details
Gene Symbol: LOC102547542 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771373.1 | XP_008769595.1 | 1 | 2966 |
Gene Symbol: LOC689574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195503.1 | NP_001182432.1 | 3 | 224 | ||
NM_001195504.1 | NP_001182433.1 | 3 | 191 | |||
GenBank mRNA | FM049367.1 | - | 3 | 113 | ||
FM067978.1 | - | 3 | 146 |
