siRNA Details
Gene Symbol: Krtcap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109674.1 | NP_001103144.1 | 4 | 504 | ||
GenBank mRNA | BC100633.1 | - | 4 | 451 | 5622000 |

More Information
Other Species Targeted

