siRNA Details
Gene Symbol: Myeov2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109044.1 | NP_001102514.1 | 3 | 366 | ||
GenBank mRNA | BC169124.1 | - | 3 | 369 | 6890894 | |
DV717666.1 | - | 3 | 366 | |||
FQ212403.1 | - | 3 | 358 | |||
FQ221993.1 | - | 3 | 357 | |||
FQ223515.1 | - | 3 | 357 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
