siRNA Details
Gene Symbol: Ankmy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006245554.2 | XP_006245616.1 | 9 | 2569 | ||
XM_008767387.1 | XP_008765609.1 | 8 | 2813 | |||
XM_008767388.1 | XP_008765610.1 | 8 | 2813 | |||
XM_008767389.1 | XP_008765611.1 | 8 | 2813 | |||
XM_008767390.1 | XP_008765612.1 | 8 | 5418 | |||
XM_008767391.1 | XP_008765613.1 | 8 | 5418 | |||
XM_008767392.1 | XP_008765614.1 | 8 | 5418 | |||
XM_008767393.1 | XP_008765615.1 | 8 | 5418 |
