siRNA Details
Gene Symbol: Cryga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080936.1 | NP_001074405.1 | 3 | 304 | ||
XM_006245063.2 | XP_006245125.1 | 9 | 1335 | |||
XM_006245064.2 | XP_006245126.1 | 9 | 1112 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
