siRNA Details
Gene Symbol: LRRTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109374.1 | NP_001102844.1 | 2 | 1372 | ||
XM_006236700.2 | XP_006236762.1 | 2 | 1574 |

More Information
Other Species Targeted

Gene Ontology Categories:
