siRNA Details
Gene Symbol: Fam91a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127578.1 | NP_001121050.1 | 18 | 1979 | ||
XM_006241684.2 | XP_006241746.1 | 18 | 1983 | |||
GenBank mRNA | BC166852.1 | - | 18 | 1979 | 9092514 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
