siRNA Details
Gene Symbol: Ackr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006243689.2 | XP_006243751.1 | 2 | 1108 | ||
XM_008766591.1 | XP_008764813.1 | 2 | 1478 |

More Information
Other Species Targeted

Gene Ontology Categories:
