siRNA Details
Gene Symbol: Dda1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134790.1 | NP_001128262.1 | 3, 4 | 247 | ||
XM_006252948.2 | XP_006253010.1 | 3, 4 | 1713 | |||
GenBank mRNA | BF549138.1 | - | 3, 4 | 260 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
