siRNA Details
Gene Symbol: Sik3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766173.1 | XP_008764395.1 | 22 | 3854 | ||
XM_008766174.1 | XP_008764396.1 | 21 | 3709 | |||
XM_008766175.1 | XP_008764397.1 | 21 | 3709 | |||
GenBank mRNA | BC166518.1 | - | 3 | 1364 |

More Information
Other Species Targeted

Gene Ontology Categories:
