siRNA Details
Gene Symbol: Sik3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766173.1 | XP_008764395.1 | 20, 21 | 2812 | ||
XM_008766174.1 | XP_008764396.1 | 19, 20 | 2667 | |||
XM_008766175.1 | XP_008764397.1 | 19, 20 | 2667 | |||
GenBank mRNA | BC166518.1 | - | 1, 2 | 322 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
