siRNA Details
Gene Symbol: LOC100911148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229637.2 | XP_006229699.1 | 3 | 538 |
Gene Symbol: LOC690082 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759620.1 | XP_008757842.1 | 1 | 1414 |
