siRNA Details
Gene Symbol: Lsm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004073.1 | NP_001004073.1 | 4, 5 | 270 | ||
NM_001165922.1 | NP_001159394.1 | 4, 5 | 367 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lsm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004073.1 | NP_001004073.1 | 4, 5 | 270 | ||
NM_001165922.1 | NP_001159394.1 | 4, 5 | 367 |