siRNA Details
Gene Symbol: Tmem221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728393.4 | XP_002728439.2 | 1, 2 | 1105 | ||
XM_006252969.2 | XP_006253031.1 | 1, 2 | 1105 |

More Information
Other Species Targeted

