siRNA Details
Gene Symbol: Klrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001072378.4 | XP_001072378.2 | 3 | 1529 | ||
XM_006236344.2 | XP_006236406.1 | 3 | 1530 | |||
XM_006236345.2 | XP_006236407.1 | 3 | 1530 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
