siRNA Details
Gene Symbol: Snrnp27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108636.1 | NP_001102106.1 | 5, 6 | 455 | ||
GenBank mRNA | FQ224589.1 | - | 457 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025665, AK011937, AK160190, BC027564.
