siRNA Details
Gene Symbol: LOC688459 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109502.1 | NP_001102972.1 | 1 | 340 | ||
GenBank mRNA | BC167028.1 | - | 1 | 324 | 9091474 | |
FQ212996.1 | - | 1 | 339 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
