siRNA Details
Gene Symbol: T2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161835.1 | NP_001155307.1 | 11 | 1229 | ||
XM_006227933.2 | XP_006227995.1 | 11 | 2191 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
