siRNA Details
Gene Symbol: Clgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109472.1 | NP_001102942.1 | 2 | 343 | ||
XM_006255298.2 | XP_006255360.1 | 2 | 296 | |||
XM_006255299.2 | XP_006255361.1 | 2 | 377 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
