siRNA Details
Gene Symbol: Glyctk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109449.2 | NP_001102919.1 | 5 | 897 | ||
XM_006243856.2 | XP_006243918.1 | 5 | 1172 | |||
XM_006243857.2 | XP_006243919.1 | 5 | 955 | |||
GenBank mRNA | BC105621.1 | - | 5 | 897 | 7454021 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
