siRNA Details
Gene Symbol: Kcmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128192.1 | NP_001121664.1 | 1 | 173 | ||
GenBank mRNA | BC167075.1 | - | 1 | 173 | 9093013 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_019715, BC043330, BC029325, AK007070, AK142436.
Gene Ontology Categories:
