siRNA Details
Gene Symbol: FAM131B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031690.2 | NP_001026860.2 | 5 | 556 | ||
NM_001278297.1 | NP_001265226.1 | 4 | 462 | |||
NM_014690.4 | NP_055505.3 | 5 | 449 | |||
XM_005250073.3 | XP_005250130.1 | 5 | 770 | |||
XM_006716186.2 | XP_006716249.1 | 5 | 548 | |||
GenBank mRNA | AB018316.1 | - | 4 | 344 | ||
AK124557.1 | - | 5 | 504 | |||
AK291470.1 | - | 5 | 421 | |||
AL832579.2 | - | 4 | 2037 | |||
BC045611.1 | - | 5 | 561 | 5300056 | ||
BC050543.1 | - | 5 | 424 | 5768309 | ||
DA334424.1 | - | 4 | 347 |
