siRNA Details
Gene Symbol: Unc93a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001061886.4 | XP_001061886.2 | 5 | 995 | ||
XM_006227938.2 | XP_006228000.1 | 5 | 826 | |||
XM_008758782.1 | XP_008757004.1 | 5 | 831 | |||
XM_008758783.1 | XP_008757005.1 | 6 | 1311 | |||
XM_008758784.1 | XP_008757006.1 | 7 | 972 | |||
XM_008758785.1 | XP_008757007.1 | 5 | 1743 | |||
XM_008758786.1 | XP_008757008.1 | 8 | 1261 | |||
XM_008758787.1 | XP_008757009.1 | 6 | 1594 | |||
XM_008758788.1 | XP_008757010.1 | 5 | 1208 | |||
XM_008758789.1 | XP_008757011.1 | 5 | 1982 | |||
XM_008758790.1 | XP_008757012.1 | 6 | 898 | |||
XM_008758791.1 | XP_008757013.1 | 5 | 537 | |||
XM_008758792.1 | XP_008757014.1 | 6 | 1062 | |||
XM_008758793.1 | XP_008757015.1 | 5 | 1972 | |||
XM_008758794.1 | XP_008757016.1 | 5 | 639 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
