siRNA Details
Gene Symbol: ECE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037324.2 | NP_001032401.1 | 4, 5 | 667 | ||
NM_001100120.1 | NP_001093590.1 | 5, 6 | 892 | |||
NM_001100121.1 | NP_001093591.1 | 5, 6 | 754 | |||
NM_014693.3 | NP_055508.3 | 5, 6 | 1003 | |||
GenBank mRNA | AB011176.1 | - | 5, 6 | 749 | ||
AF192531.1 | - | 5, 6 | 659 | |||
AF428263.1 | - | 5, 6 | 931 | |||
AF428264.1 | - | 4, 5 | 572 | |||
AF521189.1 | - | 5, 6 | 868 | |||
AK293713.1 | - | 5, 6 | 773 | |||
AK293891.1 | - | 4, 5 | 597 | |||
AK296541.1 | - | 4, 5 | 650 | |||
AK309236.1 | - | 3, 4 | 447 | |||
AY359003.1 | - | 4, 5 | 623 | |||
BC142950.1 | - | 4, 5 | 667 | 8860334 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
