siRNA Details
Gene Symbol: Cenpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750123.2 | XP_003750171.1 | 2 | 127 | ||
XM_006239855.1 | XP_006239917.1 | 2 | 127 | |||
XM_008764544.1 | XP_008762766.1 | 2 | 127 | |||
XM_008764545.1 | XP_008762767.1 | 2 | 127 | |||
XM_008764546.1 | XP_008762768.1 | 2 | 127 |
