siRNA Details
Gene Symbol: Gba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127639.1 | NP_001121111.1 | 8, 9 | 1313 | ||
GenBank mRNA | BC166732.1 | - | 8, 9 | 1313 | 7934870 |

More Information
Other Species Targeted

Gene Ontology Categories:
