siRNA Details
Gene Symbol: LOC681309 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761238.1 | XP_008759460.1 | 12, 13 | 1527 |

More Information
Other Species Targeted

Gene Ontology Categories:
