siRNA Details
Gene Symbol: Zc3h10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191090.1 | NP_001178019.1 | 3 | 353 | ||
XM_008765041.1 | XP_008763263.1 | 3 | 556 | |||
XM_008765042.1 | XP_008763264.1 | 4 | 817 | |||
XM_008765043.1 | XP_008763265.1 | 4 | 720 |

More Information
Other Species Targeted

Gene Ontology Categories:
