siRNA Details
Gene Symbol: Slc17a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271214.1 | NP_001258143.1 | 6 | 821 | ||
XM_008771657.1 | XP_008769879.1 | 6 | 826 | |||
XM_008771658.1 | XP_008769880.1 | 7 | 1140 | |||
XM_008771659.1 | XP_008769881.1 | 7 | 1210 | |||
XM_008771660.1 | XP_008769882.1 | 5 | 592 |
