siRNA Details
Gene Symbol: RAB11FIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142272.1 | NP_001135744.1 | 7 | 911 | ||
NM_014700.3 | NP_055515.1 | 9 | 1957 | |||
XM_005255713.3 | XP_005255770.1 | 10 | 2127 | |||
XM_005255714.1 | XP_005255771.1 | 8 | 972 | |||
XM_005255715.4 | XP_005255772.1 | 8 | 1778 | |||
XM_011522764.2 | XP_011521066.1 | 8 | 1004 | |||
XM_011522765.2 | XP_011521067.1 | 8 | 830 | |||
GenBank mRNA | AB014565.1 | - | 9 | 2005 | ||
AF395731.1 | - | 9 | 2005 | |||
AK293644.1 | - | 7 | 826 | |||
AK303061.1 | - | 7 | 915 | |||
AY130007.1 | - | 1 | 26 | |||
BC051360.1 | - | 9 | 1956 | 6646253 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
