siRNA Details
Gene Symbol: Slc25a42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127590.1 | NP_001121062.1 | 5 | 380 | ||
XM_006252950.2 | XP_006253012.1 | 5 | 461 | |||
XM_006252951.2 | XP_006253013.1 | 5 | 461 | |||
XM_008771155.1 | XP_008769377.1 | 5 | 978 | |||
GenBank mRNA | BC160887.1 | - | 5 | 380 | 7387602 |

More Information
Other Species Targeted

Gene Ontology Categories:
