siRNA Details
Gene Symbol: RAB11FIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142272.1 | NP_001135744.1 | 8 | 967 | ||
NM_014700.3 | NP_055515.1 | 10 | 2013 | |||
XM_005255713.3 | XP_005255770.1 | 11 | 2183 | |||
XM_005255714.1 | XP_005255771.1 | 9 | 1028 | |||
XM_005255715.4 | XP_005255772.1 | 9 | 1834 | |||
XM_011522764.2 | XP_011521066.1 | 9 | 1060 | |||
XM_011522765.2 | XP_011521067.1 | 9 | 886 | |||
GenBank mRNA | AB014565.1 | - | 10 | 2061 | ||
AF395731.1 | - | 10 | 2061 | |||
AK293644.1 | - | 8 | 882 | |||
AK303061.1 | - | 8 | 971 | |||
AY130007.1 | - | 2 | 82 | |||
BC051360.1 | - | 10 | 2012 | 6646253 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
