siRNA Details
Gene Symbol: Pou2af1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109599.1 | NP_001103069.1 | 4 | 507 | ||
XM_008766341.1 | XP_008764563.1 | 4 | 2773 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
