siRNA Details
Gene Symbol: Mansc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109603.2 | NP_001103073.1 | 3 | 603 | ||
XM_006237537.2 | XP_006237599.1 | 3 | 730 | |||
XM_006237538.2 | XP_006237600.1 | 3 | 735 |
