siRNA Details
Gene Symbol: Psrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044302.1 | NP_001037767.1 | 4 | 351 | ||
XM_006233176.2 | XP_006233238.1 | 4 | 561 | |||
XM_006233177.2 | XP_006233239.1 | 4 | 392 | |||
XM_006233178.2 | XP_006233240.1 | 4 | 561 | |||
XM_006233179.2 | XP_006233241.1 | 4 | 392 | |||
GenBank mRNA | BC105874.1 | - | 4 | 351 | 7131142 |
