siRNA Details
Gene Symbol: Tmem190 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001078306.4 | XP_001078306.2 | 2 | 1497 | ||
XM_008759018.1 | XP_008757240.1 | 2 | 1497 |

More Information
Other Species Targeted

Gene Ontology Categories:
