siRNA Details
Gene Symbol: Zfpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286936.1 | NP_001273865.1 | 3, 4 | 322 | ||
XM_006230901.2 | XP_006230963.1 | 3, 4 | 298 | |||
XM_008760175.1 | XP_008758397.1 | 3, 4 | 286 | |||
GenBank mRNA | CB717504.1 | - | 3, 4 | 322 | ||
DY546174.1 | - | 1, 2 | 42 | |||
FQ227385.1 | - | 3, 4 | 358 |

More Information
Other Species Targeted

Gene Ontology Categories:
