siRNA Details
Gene Symbol: Cdca5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115042.2 | NP_001108514.2 | 5 | 652 | ||
XM_008760176.1 | XP_008758398.1 | 5 | 647 | |||
GenBank mRNA | BC097411.1 | - | 5 | 582 | ||
BC158663.1 | - | 5 | 652 | 8369770 | ||
BC158764.1 | - | 5 | 591 | 7111047 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
