siRNA Details
Gene Symbol: Igsf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271454.1 | NP_001258383.1 | 5 | 1114 | ||
XM_008764231.1 | XP_008762453.1 | 4 | 428 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
