siRNA Details
Gene Symbol: Ttc32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750130.3 | XP_003750178.1 | 3 | 501 | ||
GenBank mRNA | FQ209446.1 | - | 2 | 225 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
