siRNA Details
Gene Symbol: Abracl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099647.1 | NP_001093117.1 | 3 | 223 | ||
GenBank mRNA | BC089921.1 | - | 1 | 456 | 7306744 | |
CO569536.1 | - | 3 | 223 | |||
FQ221434.1 | - | 3 | 286 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
